1,1,3,3-Tetramethylbutyl isocyanate, 98%, Thermo Scientific Chemicals
1,1,3,3-Tetramethylbutyl isocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1,3,3-Tetramethylbutyl isocyanate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18706.03
also known as L18706-03
Price (USD)/ Each
26.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
26.10
Add to cart
1,1,3,3-Tetramethylbutyl isocyanate, 98%, Thermo Scientific Chemicals
Catalog numberL18706.03
Price (USD)/ Each
26.10
-
Add to cart
Chemical Identifiers
CAS1611-57-0
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4260-1.4300 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only