Thermo Scientific Chemicals

2-Allyl-4,6-dibenzoylresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: L18722.06
5 g, Each
Thermo Scientific Chemicals

2-Allyl-4,6-dibenzoylresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: L18722.06
5 g, Each
Quantity
Catalog number: L18722.06
also known as L18722-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
102593-74-8
IUPAC Name
4,6-dibenzoyl-2-(prop-2-en-1-yl)benzene-1,3-diol
Molecular Formula
C23H18O4
InChI Key
FSYGSBMXRNPJAD-UHFFFAOYSA-N
SMILES
OC1=C(CC=C)C(O)=C(C=C1C(=O)C1=CC=CC=C1)C(=O)C1=CC=CC=C1
Appearance (Color)
Pale yellow to yellow or yellow-green
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5% (Non-U.S. specification)
Assay by NMR
≥97.5% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text