Thermo Scientific Chemicals

D-(-)-2-Phenylglycine methyl ester hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: L18735.22
100 g, Each
Thermo Scientific Chemicals

D-(-)-2-Phenylglycine methyl ester hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: L18735.22
100 g, Each
Quantity
Catalog number: L18735.22
also known as L18735-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19883-41-1
IUPAC Name
hydrogen methyl 2-(2-aminophenyl)acetate chloride
Molecular Formula
C9H12ClNO2
InChI Key
OZMUKUBEEKGADM-UHFFFAOYSA-N
SMILES
[H+].[Cl-].COC(=O)CC1=CC=CC=C1N
Appearance (Color)
White
Assay (Titration ex Chloride)
≥95.0 to ≤105.0%
Form
Crystals or powder or crystalline powder
Optical Rotation
-115.9 ± 0.1? (c = 1 in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text