Indium(III) chloride, anhydrous, 98+%, Thermo Scientific Chemicals
Indium(III) chloride, anhydrous, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Indium(III) chloride, anhydrous, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L18758.06
also known as L18758-06
Price (USD)/ Each
50.65
Online exclusive
55.60 
Save 4.95 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
50.65
Online exclusive
55.60 
Save 4.95 (9%)
Add to cart
Indium(III) chloride, anhydrous, 98+%, Thermo Scientific Chemicals
Catalog numberL18758.06
Price (USD)/ Each
50.65
Online exclusive
55.60 
Save 4.95 (9%)
-
Add to cart
Chemical Identifiers
CAS10025-82-8
IUPAC Nameindium(3+) trichloride
Molecular FormulaCl3In
InChI KeyPSCMQHVBLHHWTO-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[In+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Appearance (Color)White
Chloride content≥47.12%
Assay from Suppliers CofA≥98.0%
Indium(III) chloride is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst. It is an efficient catalyst for thioacetalization and its chemoselectivity. It catalyzes the enantioselective ring-opening of meso-aziridines with thiols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indium(III) chloride is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst. It is an efficient catalyst for thioacetalization and its chemoselectivity. It catalyzes the enantioselective ring-opening of meso-aziridines with thiols.

Solubility
Soluble in water and tetrahydrofuran. Sparingly soluble in alcohol and insoluble in ether.

Notes
Hygroscopic. Incompatible with strong acids and strong oxidizing agents. The molten state of indium(III) chloride may conduct electricity.
RUO – Research Use Only

General References:

  1. Mild, water tolerant Lewis acid catalyst, e.g. in allylation reactions of sensitive aldehydes with allyltributyltin: J. Org Chem., 61, 105 (1996). The allylstannane can also be generated in situ from the allyl halide in water: Tetrahedron Asym., 7, 1535 (1996). Catalyzes the Mukaiyama aldol reaction, under either solvent-free conditions: Tetrahedron Lett., 39, 1579 (1998), or in water: Chem. Commun., 1819 (1996); 861 (1998); Tetrahedron Lett., 38, 3465 (1997). For Diels-Alder reactions in water, see: Chem. Commun., 2315 (1996).
  2. Promotes efficient azidolysis of ɑß-epoxy carboxyl derivatives in water at pH 4, with results superior to Yb(OTf)3 or Sc(OTf)3: J. Org. Chem., 66, 3554 (2001).
  3. In combination with a dithiol, catalyzes the efficient transthioacetalization of acetals to dithioacetals: Synlett, 727 (2002).
  4. Aldoximes can be dehydrated to nitriles; ketoximes undergo the Beckmann rearrangement to amides: Chem. Commun., 1196 (2000).
  5. Indium enolates are readily formed by transmetallation of Li enolates. These undergo Reformatsky-type reactions with aldehydes to give ß-hydroxy esters. ɑ-Bromo enolates give Darzens-type ɑß-epoxy carbonyl compounds: J. Chem. Soc., Perkin 1, 825 (2000).
  6. Catalyst for conjugate addition of indoles to electron-deficient alkenes: Synthesis, 2165 (2001):
  7. The combination with NaBH4 has been reported to be an effective radical reducing system, replacing the environmentally unfavorable TBTH (Tri-n-butyl tin hydride, A13298), e.g. in dehalogenation and cyclization reactions: J. Am. Chem. Soc., 124, 906 (2002).
  8. In combination with Triethyl silane, A10320 and a radical initiator, generates a radical reagent, analogous to Tri-n-butyl tin hydride, A13298: Org. Lett., 6, 4981 (2004).