Thermo Scientific Chemicals

1,2-Diethylbenzene, 97%, Thermo Scientific Chemicals

Catalog number: L18775.09
10 g, Each
Thermo Scientific Chemicals

1,2-Diethylbenzene, 97%, Thermo Scientific Chemicals

Catalog number: L18775.09
10 g, Each
Quantity
Catalog number: L18775.09
also known as L18775-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
135-01-3
IUPAC Name
1,2-diethylbenzene
Molecular Formula
C10H14
InChI Key
KVNYFPKFSJIPBJ-UHFFFAOYSA-N
SMILES
CCC1=CC=CC=C1CC
Appearance (Color)
Clear colorless
Refractive Index
1.5010-1.5060 @ 20?C
Assay (GC)
≥96.0%
Form
Liquid

Description

1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.

Solubility
Miscible with alcohol, benzene and carbon tetrachloride. Immiscible with water.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text