1,2-Diethylbenzene, 97%, Thermo Scientific Chemicals
1,2-Diethylbenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Diethylbenzene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 g
10 g
Catalog number L18775.18
also known as L18775-18
Price (USD)/ Each
711.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
711.00
Add to cart
1,2-Diethylbenzene, 97%, Thermo Scientific Chemicals
Catalog numberL18775.18
Price (USD)/ Each
711.00
-
Add to cart
Chemical Identifiers
CAS135-01-3
IUPAC Name1,2-diethylbenzene
Molecular FormulaC10H14
InChI KeyKVNYFPKFSJIPBJ-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.5010-1.5060 @ 20?C
Assay (GC)≥96.0%
FormLiquid
1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diethylbenzene is used as an intermediate and a building block in organic synthesis. 1,2-Diethylbenzene is used as one of the chemical composition of essential oils from flower and leaf of korean mint.

Solubility
Miscible with alcohol, benzene and carbon tetrachloride. Immiscible with water.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Trammell, S. A.; Melde, B. J.; Zabetakis, D.; Deschamps, J. R.; Dinderman, M. A.; Johnson, B. J.; Kusterbeck, A. W. Electrochemical detection of TNT with in-line pre-concentration using imprinted diethylbenzene-bridged periodic mesoporous organosilicas. Sens. Actuators, B 2011, 155 (2), 737-744.
  2. Pizarro, C.; Suárez-Iglesias, O.; Medina, I.; Bueno, J. L. Using supercritical fluid chromatography to determine diffusion coefficients of 1,2-diethylbenzene, 1,4-diethylbenzene, 5-tert-butyl-m-xylene and phenylacetylene in supercritical carbon dioxide. J. Chromatogr. A 2007, 1167 (2), 202-209.