Thermo Scientific Chemicals

3-Ethyltoluene, 97%, Thermo Scientific Chemicals

Catalog number: L18788.14
25 g, Each
Thermo Scientific Chemicals

3-Ethyltoluene, 97%, Thermo Scientific Chemicals

Catalog number: L18788.14
25 g, Each
Quantity
Catalog number: L18788.14
also known as L18788-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
620-14-4
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Refractive Index
1.4945-1.4995 @ 20?C

Description

It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text