1,3,5-Tri-tert-butylbenzene is used in the preparation of sandwich complexes of scandium, yttrium and lanthanide ions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3,5-Tri-tert-butylbenzene is used in the preparation of sandwich complexes of scandium, yttrium and lanthanide ions.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Biedermann, M.; Grob, K. Comprehensive two-dimensional gas chromatography for characterizing mineral oils in foods and distinguishing them from synthetic hydrocarbons. J. Chromatogr. A 2015, 1375, 146-153.
Novell, A.; Minguillón, C. Monolithic silica columns with covalently attached octaproline chiral selector. Dependence of performance on derivatization degree and comparison with a bead-based analogue. J. Chromatogr. A 2015, 1384, 124-132.