Thermo Scientific Chemicals

(S)-(+)-4-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals

Catalog number: L18882.MD
250 mg, Each
Thermo Scientific Chemicals

(S)-(+)-4-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals

Catalog number: L18882.MD
250 mg, Each
Quantity
Catalog number: L18882.MD
also known as L18882-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14898-80-7
IUPAC Name
4-methylpentan-2-ol
Molecular Formula
C6H14O
InChI Key
WVYWICLMDOOCFB-UHFFFAOYNA-N
SMILES
CC(C)CC(C)O
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless
Optical Rotation
20.5? ± 0.5? (neat)
Form
Liquid
Refractive Index
1.4090-1.4120 @ 20?C

Description

It is employed in the reaction of 2-methoxyethyl acetals with allylsilanes in the presence of titanium tetrachloride: regioselective carbon-oxygen bond cleavage of the unsymmetrical acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the reaction of 2-methoxyethyl acetals with allylsilanes in the presence of titanium tetrachloride: regioselective carbon-oxygen bond cleavage of the unsymmetrical acetals.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text