Thermo Scientific Chemicals

2-(2-Aminoethoxy)ethanol, 98%, Thermo Scientific Chemicals

Catalog number: L18897.36
500 g, Each
Thermo Scientific Chemicals

2-(2-Aminoethoxy)ethanol, 98%, Thermo Scientific Chemicals

Catalog number: L18897.36
500 g, Each
Quantity
Catalog number: L18897.36
also known as L18897-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
929-06-6
IUPAC Name
2-(2-aminoethoxy)ethan-1-ol
Molecular Formula
C4H11NO2
InChI Key
GIAFURWZWWWBQT-UHFFFAOYSA-N
SMILES
NCCOCCO
Refractive Index
1.4570-1.4610 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Form
Liquid

Description

2-(2-Aminoethoxy)ethanol is used mainly in stripper solutions for applications in electronics, in gas treating to remove carbon dioxide and hydrogen sulfide, and in coolants/lubricants for metal working applications. Other applications of aminoethoxyethanol (ADG) include, inter alia, crop protection products, surfactants and colorants. It is also used as a selective solvent for recovery of aromatics from refinery streams and in preparation of foam stabilizers, wetting and emulsifying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Aminoethoxy)ethanol is used mainly in stripper solutions for applications in electronics, in gas treating to remove carbon dioxide and hydrogen sulfide, and in coolants/lubricants for metal working applications. Other applications of aminoethoxyethanol (ADG) include, inter alia, crop protection products, surfactants and colorants. It is also used as a selective solvent for recovery of aromatics from refinery streams and in preparation of foam stabilizers, wetting and emulsifying agents.

Solubility
Fully miscible in water, alcohols and aromatic hydrocarbons.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to air. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text