Thermo Scientific Chemicals

Methyl nitroacetate, 97+%, Thermo Scientific Chemicals

Catalog number: L18950.04
2 g, Each
Thermo Scientific Chemicals

Methyl nitroacetate, 97+%, Thermo Scientific Chemicals

Catalog number: L18950.04
2 g, Each
Quantity
Catalog number: L18950.04
also known as L18950-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2483-57-0
IUPAC Name
methyl 2-nitroacetate
Molecular Formula
C3H5NO4
InChI Key
ALBSWLMUHHZLLR-UHFFFAOYSA-N
SMILES
COC(=O)C[N+]([O-])=O
Assay from Suppliers CofA
≥97.5% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms [non-U.S. specification]
Refractive Index
1.4235-1.4285 @ 20°C (non-U.S. specification)
Appearance (Color)
Clear colorless to yellow

Description

Methyl nitroacetate is used in the synthesis of aromatic and heteroaromatic linear (E)-alfa-nitro-arylpentenoates. It is also used to prepare phenyliodonium ylide. Further, it is utilized in the highly enantioselective and diastereoselective copper(I)-catalyzed cyclopropanation of alkenes. In addition to this, it is employed in the production of methyl (Z)-2-nitro-3-(4-nitrophenyl)-2-propenoate through reaction with 4-nitrobenzylideneaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl nitroacetate is used in the synthesis of aromatic and heteroaromatic linear (E)-alfa-nitro-arylpentenoates. It is also used to prepare phenyliodonium ylide. Further, it is utilized in the highly enantioselective and diastereoselective copper(I)-catalyzed cyclopropanation of alkenes. In addition to this, it is employed in the production of methyl (Z)-2-nitro-3-(4-nitrophenyl)-2-propenoate through reaction with 4-nitrobenzylideneaniline.

Solubility
Miscible with ethanol, ether and most organic solvents.

Notes
Light sensitive. Incompatible withstrong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text