Thermo Scientific Chemicals

o-Toluenesulfonyl chloride, 80%, Thermo Scientific Chemicals

Catalog number: L18983.14
25 g, Each
Thermo Scientific Chemicals

o-Toluenesulfonyl chloride, 80%, Thermo Scientific Chemicals

Catalog number: L18983.14
25 g, Each
Quantity
Catalog number: L18983.14
also known as L18983-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
133-59-5
IUPAC Name
2-methylbenzene-1-sulfonyl chloride
Molecular Formula
C7H7ClO2S
InChI Key
HDECRAPHCDXMIJ-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1S(Cl)(=O)=O
Assay (GC)
≥75.0%
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Refractive Index
1.5535-1.5585 @ 20?C

Description

o-Toluenesulfonyl chloride is used in organic synthesis and saccharin production. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluenesulfonyl chloride is used in organic synthesis and saccharin production. It is also used as pharmaceutical intermediate.

Solubility
Soluble in hot alcohol, ether and benzene. Reacts with water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Store under inert gas. It is sensitive to moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text