Thermo Scientific Chemicals

D-Epoxone, 98%, Thermo Scientific Chemicals

Catalog number: L18989.03
1 g, Each
Thermo Scientific Chemicals

D-Epoxone, 98%, Thermo Scientific Chemicals

Catalog number: L18989.03
1 g, Each
Quantity
Catalog number: L18989.03
also known as L18989-03
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
18422-53-2
IUPAC Name
2,2,5',5'-tetramethyl-tetrahydro-2H-spiro[[1,3]dioxolo[4,5-c]pyran-6,2'-[1,4]dioxolan]-7-one
Molecular Formula
C12H18O6
InChI Key
IVWWFWFVSWOTLP-UHFFFAOYNA-N
SMILES
CC1(C)OC2COC3(COC(C)(C)O3)C(=O)C2O1
Appearance (Color)
White
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
97-106°C
Optical Rotation
-122.0 ± 1.5° (c=1 in chloroform)

Description

D-Epoxone is a chemical reagent used for enantioselective epoxidation of alkenes. It is involved in the synthesis of Irciniastatin (A and B), which are used as antitumor compounds. It is used as cytotoxic secondary metabolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Epoxone is a chemical reagent used for enantioselective epoxidation of alkenes. It is involved in the synthesis of Irciniastatin (A and B), which are used as antitumor compounds. It is used as cytotoxic secondary metabolite.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text