D-Epoxone, 98%, Thermo Scientific Chemicals
D-Epoxone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Epoxone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L18989.06
also known as L18989-06
Price (USD)/ Each
213.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
213.00
Add to cart
D-Epoxone, 98%, Thermo Scientific Chemicals
Catalog numberL18989.06
Price (USD)/ Each
213.00
-
Add to cart
Chemical Identifiers
CAS18422-53-2
IUPAC Name2,2,5',5'-tetramethyl-tetrahydro-2H-spiro[[1,3]dioxolo[4,5-c]pyran-6,2'-[1,4]dioxolan]-7-one
Molecular FormulaC12H18O6
InChI KeyIVWWFWFVSWOTLP-UHFFFAOYNA-N
SMILESCC1(C)OC2COC3(COC(C)(C)O3)C(=O)C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White
Identification (FTIR)Conforms
Melting Point (clear melt)97-106°C
Optical Rotation-122.0 ± 1.5° (c=1 in chloroform)
View more
D-Epoxone is a chemical reagent used for enantioselective epoxidation of alkenes. It is involved in the synthesis of Irciniastatin (A and B), which are used as antitumor compounds. It is used as cytotoxic secondary metabolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Epoxone is a chemical reagent used for enantioselective epoxidation of alkenes. It is involved in the synthesis of Irciniastatin (A and B), which are used as antitumor compounds. It is used as cytotoxic secondary metabolite.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Tran, A. T.; Luo, B.; Jagadeesh, Y.; Auberger, N.; Désiré, J.; Nakagawa, S.; Kato, A.; Zhang, Y.; Blériot, Y.; Sollogoub, M. Synthesis of pyrrolidine-based analogues of 2-acetamidosugars as N-acetyl-d-glucosaminidase inhibitors. Carbohydr. Res. 2015, 409, 56-62.
  2. Ren, X-D.; Zhao, N.; Xu, S.; Lü, H-N.; Ma, S-G.; Liu, Y-B.; Li, Y.; Qu, J.; Yu, S-S. Total synthesis of illicidione A and illihendione A. Tetrahedron 2015, 71 (29), 4821-4829.