Thermo Scientific Chemicals

(R)-(+)-3-Hydroxy-3-phenylpropionic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L19040.MD
250 mg, Each
Thermo Scientific Chemicals

(R)-(+)-3-Hydroxy-3-phenylpropionic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L19040.MD
250 mg, Each
Quantity
Catalog number: L19040.MD
also known as L19040-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2768-42-5
IUPAC Name
(3R)-3-hydroxy-3-phenylpropanoate
Molecular Formula
C9H9O3
InChI Key
AYOLELPCNDVZKZ-MRVPVSSYSA-M
SMILES
O[C@H](CC([O-])=O)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White to pale cream
Assay (Silylated GC)
≥98.0%
Optical Rotation
22.2 ± 0.5? (C=4 in methanol)
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

It is employed in the synthesis and evaluation of a novel, reusable solid-supported open chain chiral auxiliary derived from m-hydrobenzoin for the diastereoselective reduction of α-keto esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis and evaluation of a novel, reusable solid-supported open chain chiral auxiliary derived from m-hydrobenzoin for the diastereoselective reduction of α-keto esters.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text