Thermo Scientific Chemicals

(R)-(-)-1-Cyclohexylethylamine, ChiPros 98%, ee 94+%, Thermo Scientific Chemicals

Catalog number: L19051.03
1 g, Each
Thermo Scientific Chemicals

(R)-(-)-1-Cyclohexylethylamine, ChiPros 98%, ee 94+%, Thermo Scientific Chemicals

Catalog number: L19051.03
1 g, Each
Quantity
Catalog number: L19051.03
also known as L19051-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5913-13-3
Appearance (Color)
Clear, colourless
Assay (GC)
>97.5%
Refractive Index
1.4600-1.4640
Optical Rotation
-4? ?0.5? (neat)
Form
liquid

Description

(R)-(-)-1-Cyclohexylethylamine is used in the preparation of benzylidene(1-cyclohexylethyl)amine by reacting with benzaldehyde. It is also used to prepare chiral polymethacrylamides, which are employed as stationary phases for the liquid-chromatographic resolution of racemic amides. In addition to this, it is used in the preparation of polyamides bound to silica gel for high performance liquid chromatography, which are pressure-stable.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1-Cyclohexylethylamine is used in the preparation of benzylidene(1-cyclohexylethyl)amine by reacting with benzaldehyde. It is also used to prepare chiral polymethacrylamides, which are employed as stationary phases for the liquid-chromatographic resolution of racemic amides. In addition to this, it is used in the preparation of polyamides bound to silica gel for high performance liquid chromatography, which are pressure-stable.

Solubility
Miscible with acetone, chloroform,dimethyl sulfoxide, ethanol, methanol, ether, petroleum ether and dichloromethane. Insoluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text