4-Methoxyindole, 99%, Thermo Scientific Chemicals
4-Methoxyindole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxyindole, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number L19069.03
also known as L19069-03
Price (USD)/ Each
165.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
165.00
Add to cart
4-Methoxyindole, 99%, Thermo Scientific Chemicals
Catalog numberL19069.03
Price (USD)/ Each
165.00
-
Add to cart
Chemical Identifiers
CAS4837-90-5
IUPAC Name4-methoxy-1H-indole
Molecular FormulaC9H9NO
InChI KeyLUNOXNMCFPFPMO-UHFFFAOYSA-N
SMILESCOC1=C2C=CNC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)67.5-71.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only