5-Methoxyindole-3-acetic acid, 98+%, Thermo Scientific Chemicals
5-Methoxyindole-3-acetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Methoxyindole-3-acetic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19073.MD
also known as L19073-MD
Price (USD)/ Each
53.10
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
53.10
Add to cart
5-Methoxyindole-3-acetic acid, 98+%, Thermo Scientific Chemicals
Catalog numberL19073.MD
Price (USD)/ Each
53.10
-
Add to cart
Chemical Identifiers
CAS3471-31-6
IUPAC Name2-(5-methoxy-1H-indol-3-yl)acetate
Molecular FormulaC11H10NO3
InChI KeyCOCNDHOPIHDTHK-UHFFFAOYSA-M
SMILESCOC1=CC=C2NC=C(CC([O-])=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (Silylated GC)≥98.0%
Appearance (Color)White to cream
Melting Point (clear melt)145.0-152.0?C
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only