Benzenesulfonic acid, tech. ca 75% w/w aq. soln., Thermo Scientific Chemicals
Benzenesulfonic acid, tech. ca 75% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzenesulfonic acid, tech. ca 75% w/w aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
5000 mL
Catalog number L19077.AK
also known as L19077-AK
Price (USD)/ Each
24.50
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
24.50
Add to cart
Benzenesulfonic acid, tech. ca 75% w/w aq. soln., Thermo Scientific Chemicals
Catalog numberL19077.AK
Price (USD)/ Each
24.50
-
Add to cart
Chemical Identifiers
CAS98-11-3
IUPAC Namebenzenesulfonic acid
Molecular FormulaC6H6O3S
InChI KeySRSXLGNVWSONIS-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Appearance (Color)Colourless to dark brown
Assay (Aqueous acid-base Titration)> 73.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only