Thermo Scientific Chemicals

Benzenesulfonic acid, tech. ca 75% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: L19077.AU
1000 mL, Each
Thermo Scientific Chemicals

Benzenesulfonic acid, tech. ca 75% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: L19077.AU
1000 mL, Each
Quantity
Catalog number: L19077.AU
also known as L19077-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-11-3
IUPAC Name
benzenesulfonic acid
Molecular Formula
C6H6O3S
InChI Key
SRSXLGNVWSONIS-UHFFFAOYSA-N
SMILES
OS(=O)(=O)C1=CC=CC=C1
Form
liquid
Appearance (Color)
Colourless to dark brown
Assay (Aqueous acid-base Titration)
> 73.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text