1-Amino-4-methylpiperazine, 98%, Thermo Scientific Chemicals
1-Amino-4-methylpiperazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Amino-4-methylpiperazine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number L19108.22
also known as L19108-22
Price (USD)/ Each
148.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
148.00
Add to cart
1-Amino-4-methylpiperazine, 98%, Thermo Scientific Chemicals
Catalog numberL19108.22
Price (USD)/ Each
148.00
-
Add to cart
Chemical Identifiers
CAS6928-85-4
IUPAC Name4-methylpiperazin-1-amine
Molecular FormulaC5H13N3
InChI KeyRJWLLQWLBMJCFD-UHFFFAOYSA-N
SMILESCN1CCN(N)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4825-1.4865 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only