Thermo Scientific Chemicals

(R)-(+)-1-Phenylethylamine, 99+%, ee 99+%, Thermo Scientific Chemicals

Catalog number: L19117.14
25 g, Each
Thermo Scientific Chemicals

(R)-(+)-1-Phenylethylamine, 99+%, ee 99+%, Thermo Scientific Chemicals

Catalog number: L19117.14
25 g, Each
Quantity
Catalog number: L19117.14
also known as L19117-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3886-69-9
IUPAC Name
1-phenylethan-1-amine
Molecular Formula
C8H11N
InChI Key
RQEUFEKYXDPUSK-UHFFFAOYNA-N
SMILES
CC(N)C1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥99.0%
Optical Rotation
+30° ± 2° (c=10 in ethanol)
Refractive Index
1.5250-1.5280 @ 20°C

Description

(R)-(+)-1-Phenylethylamine is used as an intermediate in organic synthesis. It is also used to determine the enantiomeric purity of acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-Phenylethylamine is used as an intermediate in organic synthesis. It is also used to determine the enantiomeric purity of acids.

Solubility
Miscible with water and chloroform.

Notes
Store in a cool place. Air Sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text