2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- M. Jonathan Fray, Kelvin Cooper, M. John Parry, Kenneth Richardson, John Steele. Novel Antagonists of Platelet-Activating Factor. 1. Synthesis and Structure-Activity Relationships of Benzodiazepine and Benzazepine Derivatives of 2-Methyl-1-phenylimidazo[4,5-c]pyridine. J. Med. Chem. 1995, 38 (18), 3514-3523.
- M. Jonathan Fray, David J. Bull, Kelvin Cooper, M. John Parry, Mark H. Stefaniak. Novel Antagonists of Platelet-Activating Factor. 2. Synthesis and Structure-Activity Relationships of Potent and Long-Acting Heterofused [1,5]Benzodiazepine and [1,4]Diazepine Derivatives of 1-Phenyl-2-methylimidazo[4,5-c]pyridine. J. Med. Chem. 1995, 38 (18), 3524-3535.