Thermo Scientific Chemicals

2-Methylimidazo[4,5-c]pyridine, 96%, Thermo Scientific Chemicals

Catalog number: L19131.MD
250 mg, Each
Thermo Scientific Chemicals

2-Methylimidazo[4,5-c]pyridine, 96%, Thermo Scientific Chemicals

Catalog number: L19131.MD
250 mg, Each
Quantity
Catalog number: L19131.MD
also known as L19131-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63604-59-1
IUPAC Name
2-methyl-3H-imidazo[4,5-c]pyridine
Molecular Formula
C7H7N3
InChI Key
JWQVOMJORQXNME-UHFFFAOYSA-N
SMILES
CC1=NC2=CC=NC=C2N1
Appearance (Color)
White to cream or yellow to pale brown
Assay (Non-aqueous acid-base Titration)
≥95.0% to ≤105.0%
Form
Powder
Melting Point (clear melt)
169-179?C

Description

2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text