2,2,3,3-Tetrafluorocyclobutanol, 95%, Thermo Scientific Chemicals
2,2,3,3-Tetrafluorocyclobutanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,3,3-Tetrafluorocyclobutanol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19186.03
also known as L19186-03
Price (USD)/ Each
154.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
154.00
Add to cart
2,2,3,3-Tetrafluorocyclobutanol, 95%, Thermo Scientific Chemicals
Catalog numberL19186.03
Price (USD)/ Each
154.00
-
Add to cart
Chemical Identifiers
CAS374-32-3
IUPAC Name2,2,3,3-tetrafluorocyclobutan-1-ol
Molecular FormulaC4H4F4O
InChI KeyFNENUTDUAXPVTF-UHFFFAOYNA-N
SMILESOC1CC(F)(F)C1(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Refractive Index1.3460-1.3520 @ 20°C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
2,2,3,3-Tetrafluorocyclobutanol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-Tetrafluorocyclobutanol is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only