Thermo Scientific Chemicals

2,2,3,3-Tetrafluorocyclobutanol, 95%, Thermo Scientific Chemicals

Catalog number: L19186.06
5 g, Each
Thermo Scientific Chemicals

2,2,3,3-Tetrafluorocyclobutanol, 95%, Thermo Scientific Chemicals

Catalog number: L19186.06
5 g, Each
Quantity
Catalog number: L19186.06
also known as L19186-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
374-32-3
IUPAC Name
2,2,3,3-tetrafluorocyclobutan-1-ol
Molecular Formula
C4H4F4O
InChI Key
FNENUTDUAXPVTF-UHFFFAOYNA-N
SMILES
OC1CC(F)(F)C1(F)F
Assay (GC)
≥94.0%
Refractive Index
1.3460-1.3520 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

2,2,3,3-Tetrafluorocyclobutanol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-Tetrafluorocyclobutanol is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text