Thermo Scientific Chemicals

Ethyl glyoxylate, ca 50% soln. in toluene, Thermo Scientific Chemicals

Catalog number: L19207.22
100 g, Each
Thermo Scientific Chemicals

Ethyl glyoxylate, ca 50% soln. in toluene, Thermo Scientific Chemicals

Catalog number: L19207.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L19207.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
924-44-7
IUPAC Name
ethyl 2-oxoacetate
Molecular Formula
C4H6O3
InChI Key
DBPFRRFGLYGEJI-UHFFFAOYSA-N
SMILES
CCOC(=O)C=O
Form
Solution
Comment
Exists partly in the polymerised form; depolymerises on heating
Appearance (Color)
Clear colorless to pale yellow
Acid-base back titration
≥45.0% to ≤55.0% (Aldehyde function)

Description

Ethyl glyoxylate is widely used as an intermediate in the synthesis of pharmaceuticals (high reactivity of aldehyde function). It is used in the synthesis of a biodegradable polymer, poly(ethyl glyoxylate). It is actively involved in the Friedel-Crafts alkylation reactions with thiophenes to give the corresponding secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl glyoxylate is widely used as an intermediate in the synthesis of pharmaceuticals (high reactivity of aldehyde function). It is used in the synthesis of a biodegradable polymer, poly(ethyl glyoxylate). It is actively involved in the Friedel-Crafts alkylation reactions with thiophenes to give the corresponding secondary alcohols.

Solubility
Immiscible with water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text