Ethyl 4-(bromomethyl)benzoate, 96%, Thermo Scientific Chemicals
Ethyl 4-(bromomethyl)benzoate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4-(bromomethyl)benzoate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number L19219.22
also known as L19219-22
Price (USD)/ Each
499.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
499.00
Add to cart
Ethyl 4-(bromomethyl)benzoate, 96%, Thermo Scientific Chemicals
Catalog numberL19219.22
Price (USD)/ Each
499.00
-
Add to cart
Chemical Identifiers
CAS26496-94-6
IUPAC Nameethyl 4-(bromomethyl)benzoate
Molecular FormulaC10H11BrO2
InChI KeyTWQLMAJROCNXEA-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
Melting Point (clear melt)28-40?C
FormCrystals or fused solid and/or lump
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only