Thermo Scientific Chemicals

Tetrathiafulvalene, 97%, Thermo Scientific Chemicals

Catalog number: L19229.MD
250 mg, Each
Thermo Scientific Chemicals

Tetrathiafulvalene, 97%, Thermo Scientific Chemicals

Catalog number: L19229.MD
250 mg, Each
Quantity
Catalog number: L19229.MD
also known as L19229-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
31366-25-3
IUPAC Name
2-(2H-1,3-dithiol-2-ylidene)-2H-1,3-dithiole
Molecular Formula
C6H4S4
InChI Key
FHCPAXDKURNIOZ-UHFFFAOYSA-N
SMILES
S1C=CSC1=C1SC=CS1
Melting Point (clear melt)
115.0-124.0?C
Appearance (Color)
Orange to brown
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

Tetrathiafulvalene, holds wide application in HPLC, NMR. This heterocyclic compound contributed to the development of molecular electronics. They are used as a organic super conductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrathiafulvalene, holds wide application in HPLC, NMR. This heterocyclic compound contributed to the development of molecular electronics. They are used as a organic super conductors.

Solubility
It is insoluble in water. Soluble in organic solvents.

Notes
Air & Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text