2-Fluoro-5-nitrobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Fluoro-5-nitrobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
It is insoluble in water.
Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
General References:
- DJ Gale.; JFK Wilshire. The preparation of some polymethine astrazon dyes. Australian Journal of Chemistry. 197023 (5), 1063 - 1068.
- X Ouyang.; AS Kiselyov. Novel synthesis of dibenzo [b, g] 1, 5-oxazocines. Tetrahedron letters. 199940 (32), 5827-5830.