3-Benzyloxyphenyl isothiocyanate, 97%, Thermo Scientific Chemicals
3-Benzyloxyphenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Benzyloxyphenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number L19242.03
also known as L19242-03
Price (USD)/ Each
126.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
126.00
Add to cart
3-Benzyloxyphenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Catalog numberL19242.03
Price (USD)/ Each
126.00
-
Add to cart
Specifications
Chemical Name or Material3-Benzyloxyphenyl isothiocyanate
CAS206559-36-6
Health Hazard 1H302+H312+H332-H314-H335
Health Hazard 2GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only