4-Iodobenzyl alcohol, 97%, Thermo Scientific Chemicals
4-Iodobenzyl alcohol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Iodobenzyl alcohol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L19278.06
also known as L19278-06
Price (USD)/ Each
129.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
129.00
Add to cart
4-Iodobenzyl alcohol, 97%, Thermo Scientific Chemicals
Catalog numberL19278.06
Price (USD)/ Each
129.00
-
Add to cart
Chemical Identifiers
CAS18282-51-4
IUPAC Name(4-iodophenyl)methanol
Molecular FormulaC7H7IO
InChI KeyCNQRHSZYVFYOIE-UHFFFAOYSA-N
SMILESOCC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)67.0-77.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only