Thermo Scientific Chemicals

5-Aminomethyl-2-chloropyridine, 97%, Thermo Scientific Chemicals

Catalog number: L19283.03
1 g, Each
Thermo Scientific Chemicals

5-Aminomethyl-2-chloropyridine, 97%, Thermo Scientific Chemicals

Catalog number: L19283.03
1 g, Each
Quantity
Catalog number: L19283.03
also known as L19283-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97004-04-1
IUPAC Name
1-(6-chloropyridin-3-yl)methanamine
Molecular Formula
C6H7ClN2
InChI Key
XPARFBOWIYMLMY-UHFFFAOYSA-N
SMILES
NCC1=CN=C(Cl)C=C1
Appearance (Color)
White to yellow or pale cream to pale brown
Assay (GC)
≥96.0%
Melting Point
25-33?C (clear to cloudy melt)
Form
Powder or Fused solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text