Thermo Scientific Chemicals

1,2-Epoxy-3-methylbutane, 98+%, Thermo Scientific Chemicals

Catalog number: L19291.03
1 g, Each
Thermo Scientific Chemicals

1,2-Epoxy-3-methylbutane, 98+%, Thermo Scientific Chemicals

Catalog number: L19291.03
1 g, Each
Quantity
Catalog number: L19291.03
also known as L19291-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1438-14-8
IUPAC Name
(2S)-2-(propan-2-yl)oxirane
Molecular Formula
C5H10O
InChI Key
REYZXWIIUPKFTI-RXMQYKEDSA-N
SMILES
CC(C)[C@H]1CO1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.3895-1.3935 @ 20?C

Description

1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-3-methylbutane is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Store under inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text