Thermo Scientific Chemicals

Sodium 4-ethynylbenzoate, 97%, Thermo Scientific Chemicals

Catalog number: L19292.06
5 g, Each
Thermo Scientific Chemicals

Sodium 4-ethynylbenzoate, 97%, Thermo Scientific Chemicals

Catalog number: L19292.06
5 g, Each
Quantity
Catalog number: L19292.06
also known as L19292-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
144693-65-2
IUPAC Name
sodium 4-ethynylbenzoate
Molecular Formula
C9H5NaO2
InChI Key
VXBLAFDQGACQAR-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)C1=CC=C(C=C1)C#C
Appearance (Color)
Yellow to cream to orange to pale brown
Water Content (Karl Fischer Titration)
≤2.0%
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text