Thermo Scientific Chemicals

(R)-(+)-1-Phenylethanol, 99%, ee 97+%, Thermo Scientific Chemicals

Catalog number: L19296.06
5 g, Each
Thermo Scientific Chemicals

(R)-(+)-1-Phenylethanol, 99%, ee 97+%, Thermo Scientific Chemicals

Catalog number: L19296.06
5 g, Each
Quantity
Catalog number: L19296.06
also known as L19296-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1517-69-7
IUPAC Name
1-phenylethan-1-ol
Molecular Formula
C8H10O
InChI Key
WAPNOHKVXSQRPX-UHFFFAOYNA-N
SMILES
CC(O)C1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Optical Rotation
+45° ± 1° (c=5, methanol)
Form
Liquid
Enantiomeric excess
≥97.0%

Description

(R)-(+)-1-Phenylethanol is used in the synthesis of optically active products. It is also useful in the determination of enantiomeric purity and for resolutions of acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-Phenylethanol is used in the synthesis of optically active products. It is also useful in the determination of enantiomeric purity and for resolutions of acids.

Solubility
Miscible with organic solvents and slightly miscible with water.

Notes
Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text