Thermo Scientific Chemicals

3-Amino-2-naphthoic acid, 97%, Thermo Scientific Chemicals

Catalog number: L19312.06
5 g, Each
Thermo Scientific Chemicals

3-Amino-2-naphthoic acid, 97%, Thermo Scientific Chemicals

Catalog number: L19312.06
5 g, Each
Quantity
Catalog number: L19312.06
also known as L19312-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5959-52-4
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%
Form
Powder
Appearance (Color)
Yellow or yellow-brown

Description

3-Amino-2-naphthoic acid is a dyes analytical reagent which is used as an intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-2-naphthoic acid is a dyes analytical reagent which is used as an intermediate for organic synthesis.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text