Thermo Scientific Chemicals

2-Bromo-4-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: L19321.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-4-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: L19321.06
5 g, Each
Quantity
Catalog number: L19321.06
also known as L19321-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1006-41-3
IUPAC Name
2-bromo-4-fluorobenzoate
Molecular Formula
C7H3BrFO2
InChI Key
RRKPMLZRLKTDQV-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CC=C(F)C=C1Br
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
169.0-176.0?C

Description

2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.

Solubility
Soluble in water and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text