N-Boc-L-isoleucinol, 95%, Thermo Scientific Chemicals
N-Boc-L-isoleucinol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-isoleucinol, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number L19351.06
also known as L19351-06
Price (USD)/ Each
267.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
267.00
Add to cart
N-Boc-L-isoleucinol, 95%, Thermo Scientific Chemicals
Catalog numberL19351.06
Price (USD)/ Each
267.00
-
Add to cart
Chemical Identifiers
CAS106946-74-1
IUPAC Nametert-butyl N-[(2S,3S)-1-hydroxy-3-methylpentan-2-yl]carbamate
Molecular FormulaC11H23NO3
InChI KeyBPLDQMXXYMKQPW-DTWKUNHWSA-N
SMILESCC[C@H](C)[C@@H](CO)NC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
FormViscous liquid
Refractive Index1.4500-1.4560 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only