1-Formyl-4-methylpiperazine, 98%, Thermo Scientific Chemicals
1-Formyl-4-methylpiperazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Formyl-4-methylpiperazine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19358.06
also known as L19358-06
Price (USD)/ Each
43.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
43.80
Add to cart
1-Formyl-4-methylpiperazine, 98%, Thermo Scientific Chemicals
Catalog numberL19358.06
Price (USD)/ Each
43.80
-
Add to cart
Chemical Identifiers
CAS7556-55-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Refractive Index1.4890-1.4930 @ 20?C
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only