Thermo Scientific Chemicals

N-Boc-L-pipecolinic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L19361.03
1 g, Each
Thermo Scientific Chemicals

N-Boc-L-pipecolinic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L19361.03
1 g, Each
Quantity
Catalog number: L19361.03
also known as L19361-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26250-84-0
IUPAC Name
(2S)-1-[(tert-butoxy)carbonyl]piperidine-2-carboxylic acid
Molecular Formula
C11H19NO4
InChI Key
JQAOHGMPAAWWQO-QMMMGPOBSA-N
SMILES
CC(C)(C)OC(=O)N1CCCC[C@H]1C(O)=O
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder or flakes
Melting Point (clear melt)
119.0-126.0?C
Appearance (Color)
White
Optical Rotation
-46.0 ± 1.0? (C=1 in DMF)

Description

N-Boc-L-pipecolinic acid is a useful boc protected Pipecolic Acid for proteomics research in modulating ion channel activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-pipecolinic acid is a useful boc protected Pipecolic Acid for proteomics research in modulating ion channel activity.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text