Thermo Scientific Chemicals

2-Hydroxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L19400.06
5 g, Each
Thermo Scientific Chemicals

2-Hydroxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L19400.06
5 g, Each
Quantity
Catalog number: L19400.06
also known as L19400-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89466-08-0
IUPAC Name
(2-hydroxyphenyl)boronic acid
Molecular Formula
C6H7BO3
InChI Key
YDMRDHQUQIVWBE-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC=C1O
Form
Powder
Assay (HPLC)
≥96.0%
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥96.0%
Proton NMR
Conforms to structure

Description

2-Hydroxybenzeneboronic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzeneboronic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text