Thermo Scientific Chemicals

Ethyl imidazole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: L19405.03
1 g, Each
Thermo Scientific Chemicals

Ethyl imidazole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: L19405.03
1 g, Each
Quantity
Catalog number: L19405.03
also known as L19405-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33543-78-1
IUPAC Name
ethyl 1H-imidazole-2-carboxylate
Molecular Formula
C6H8N2O2
InChI Key
UHYNYIGCGVDBTC-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=NC=CN1
Form
Crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
174-183?C
Appearance (Color)
White to yellow or pale cream
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text