Ethyl imidazole-2-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl imidazole-2-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl imidazole-2-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L19405.06
also known as L19405-06
Price (USD)/ Each
305.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
305.00
Add to cart
Ethyl imidazole-2-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberL19405.06
Price (USD)/ Each
305.00
-
Add to cart
Chemical Identifiers
CAS33543-78-1
IUPAC Nameethyl 1H-imidazole-2-carboxylate
Molecular FormulaC6H8N2O2
InChI KeyUHYNYIGCGVDBTC-UHFFFAOYSA-N
SMILESCCOC(=O)C1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)174-183?C
Appearance (Color)White to yellow or pale cream
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only