(R)-(+)-Tetrahydro-2-furoic acid, 98+%, ee 98%, Thermo Scientific Chemicals
(R)-(+)-Tetrahydro-2-furoic acid, 98+%, ee 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(+)-Tetrahydro-2-furoic acid, 98+%, ee 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number L19410.06
also known as L19410-06
Price (USD)/ Each
120.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
120.00
Add to cart
(R)-(+)-Tetrahydro-2-furoic acid, 98+%, ee 98%, Thermo Scientific Chemicals
Catalog numberL19410.06
Price (USD)/ Each
120.00
-
Add to cart
Chemical Identifiers
CAS87392-05-0
IUPAC Nameoxolane-2-carboxylic acid
Molecular FormulaC5H8O3
InChI KeyUJJLJRQIPMGXEZ-UHFFFAOYNA-N
SMILESOC(=O)C1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Assay (Silylated GC)>98.0%
Refractive Index1.4580 - 1.4620
Appearance (Color)Clear, colourless
Assay (Aqueous acid-base Titration)>98.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only