Thermo Scientific Chemicals

(R)-(+)-Tetrahydro-2-furoic acid, 98+%, ee 98%, Thermo Scientific Chemicals

Catalog number: L19410.14
25 g, Each
Thermo Scientific Chemicals

(R)-(+)-Tetrahydro-2-furoic acid, 98+%, ee 98%, Thermo Scientific Chemicals

Catalog number: L19410.14
25 g, Each
Quantity
Catalog number: L19410.14
also known as L19410-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87392-05-0
IUPAC Name
oxolane-2-carboxylic acid
Molecular Formula
C5H8O3
InChI Key
UJJLJRQIPMGXEZ-UHFFFAOYNA-N
SMILES
OC(=O)C1CCCO1
Form
liquid
Assay (Silylated GC)
>98.0%
Refractive Index
1.4580 - 1.4620
Appearance (Color)
Clear, colourless
Assay (Aqueous acid-base Titration)
>98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text