Thermo Scientific Chemicals

1,3-Benzenedithiol, 97%, Thermo Scientific Chemicals

Catalog number: L19439.03
1 g, Each
Thermo Scientific Chemicals

1,3-Benzenedithiol, 97%, Thermo Scientific Chemicals

Catalog number: L19439.03
1 g, Each
Quantity
Catalog number: L19439.03
also known as L19439-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
626-04-0
IUPAC Name
benzene-1,3-dithiol
Molecular Formula
C6H6S2
InChI Key
ZWOASCVFHSYHOB-UHFFFAOYSA-N
SMILES
SC1=CC(S)=CC=C1
Appearance (Color)
Clear, colorless to pale yellow
Assay (GC)
>96.0%
Refractive Index
1.6615-1.6675 @ 20?C
Form
Liquid

Description

1,3-Benzenedithiol is used to improve power conversion efficiency in cadmium selenide nanorods/poly(3-hexylthiophene) hybrid solar cells. It acts as an intermediate as well as involved in the preparation of 1,3-bis(-styrylthio)-benzol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedithiol is used to improve power conversion efficiency in cadmium selenide nanorods/poly(3-hexylthiophene) hybrid solar cells. It acts as an intermediate as well as involved in the preparation of 1,3-bis(-styrylthio)-benzol.

Solubility
Miscible with acetonitrile.

Notes
Incompatible with strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text