Thermo Scientific Chemicals

1,4-Benzenedithiol, 97%, Thermo Scientific Chemicals

Catalog number: L19440.06
5 g, Each
Thermo Scientific Chemicals

1,4-Benzenedithiol, 97%, Thermo Scientific Chemicals

Catalog number: L19440.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L19440.06
also known as L19440-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
624-39-5
IUPAC Name
benzene-1,4-dithiol
Molecular Formula
C6H6S2
InChI Key
WYLQRHZSKIDFEP-UHFFFAOYSA-N
SMILES
SC1=CC=C(S)C=C1
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
93.0-102.0?C

Description

1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text