3,5-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
3,5-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19447.MD
also known as L19447-MD
Price (USD)/ Each
95.80
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
95.80
Add to cart
3,5-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Catalog numberL19447.MD
Price (USD)/ Each
95.80
-
Add to cart
Chemical Identifiers
CAS20857-47-0
IUPAC Name3,5-bis(trifluoromethyl)pyridine
Molecular FormulaC7H3F6N
InChI KeyRQEOYYWUVYZZLL-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(=CN=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)31.0-41.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only