Thermo Scientific Chemicals

3,4,5,6-Tetrachloropyridine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: L19450.06
5 g, Each
Thermo Scientific Chemicals

3,4,5,6-Tetrachloropyridine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: L19450.06
5 g, Each
Quantity
Catalog number: L19450.06
also known as L19450-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10469-09-7
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White
Assay (Silylated GC)
≥97.5%

Description

3,4,5,6-Tetrachloropyridine-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5,6-Tetrachloropyridine-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

Solubility
Soluble in DMSO, Methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text